Magnesium ≥99.8% (metals basis), powder -20+100 mesh

Supplier: Thermo Scientific Chemicals
Danger

000869-30 000869-A1
AA00869-30EA 80.51 USD
AA00869-30 AA00869-A1
Magnesium ≥99.8% (metals basis), powder -20+100 mesh
Magnesium

MDL: MFCD00085308 Fieser: 1,627 13,170 15,194 16,198 18,224 19,205 20,229 21,258


Purity: 99.8% CAS No.: 7439-95-4 Appearance: Silver grey Odor: Odorless UN No.: 1418 MDL No.: MFCD00085308

Formula: Mg
Boiling Pt: 1107 °C (1013 hPa)
Melting Pt: 651 °C
Density: 1,74 g/cm³ (25 °C)
Flash Pt: 500 °C
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1418
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR