Zirconium ≥95%, powder APS 2-3 µm

Supplier: Thermo Scientific Chemicals
Danger

000847-14 000847-22 000847-36
AA00847-14EA 88.4 USD
AA00847-14 AA00847-22 AA00847-36
Zirconium ≥95%, powder APS 2-3 µm
Zirconium

CAS No.: 7440-67-7 Physical Form: Packaged in water Appearance: Dark grey Odor: Odorless UN No.: 1358 MDL No.: MFCD00011303

Formula: Zr
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9
UN: 1358
ADR: 4.1,II
Merck Index: 12,10306

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR