Lead 99+% (metals basis), foil 30x30 cm, Thickness 1.6 mm (0.063 in)

Supplier: Thermo Scientific Chemicals
Danger

000708-KF 000708-KJ
AA00708-KFEA 268.22 USD
AA00708-KF AA00708-KJ
Lead 99+% (metals basis), foil 30x30 cm, Thickness 1.6 mm (0.063 in)
Lead

MDL: MFCD00134050


Purity: 99+% CAS No.: 7439-92-1 Physical Form: ≈1630g/30x30cm Appearance: Grey Odor: Odorless MDL No.: MFCD00134050

Formula: Pb
Boiling Pt: 1740 °C (1013 hPa)
Melting Pt: 327 °C
Density: 11.3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR