Zirconium ≥98.5%, powder -325 mesh

Supplier: Thermo Scientific Chemicals
Danger

000418-22 000418-88 000418-14 000418-36
AA00418-22EA 329.41 USD
AA00418-22 AA00418-88 AA00418-14 AA00418-36
Zirconium ≥98.5%, powder -325 mesh
Zirconium

CAS No.: 7440-67-7 Physical Form: Packaged in water Appearance: Dark grey Odor: Odorless UN No.: 1358 MDL No.: MFCD00011303

Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9
UN: 1358
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR