Terbium ≥99.9% (REO, rare earth oxide basis), powder -40 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
000322-03
00322-03
000322-06
00322-06
AA00322-03EA
92.8
USD
CAAA00322-03
AA00322-03
CAAA00322-06
AA00322-06
Terbium ≥99.9% (REO, rare earth oxide basis), powder -40 mesh
Terbium
Formula:
Tb MW: 158.93 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011256 CAS Number: 7440-27-9 EINECS: 231-137-6 UN: 3089 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...