Copper ≥99.99% (metals basis), foil, annealed, Thickness 0.127 mm (0.005 in)

Supplier: Thermo Scientific Chemicals

000095-FI 000095-GB
AA00095-FIEA 109 USD
AA00095-FI AA00095-GB
Copper ≥99.99% (metals basis), foil, annealed, Thickness 0.127 mm (0.005 in)
Copper

MDL: MFCD00010965


Purity: 99.99+% CAS No.: 7440-50-8 Physical Form: ≈2.8g/50x50mm Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR