VWR® Hybridization Cocktail
Supplier: Avantor
VWR®
0973-50ML
97062-538EA 94.38 USD
97062-538
VWR® Hybridization Cocktail
Buffers Electrophoresis Buffers Hybridization Buffers
Offers blocking and hybridization for optimized complimentary base pairing of DNA and RNA in Southern blot, Northern blot, dot blot, colony and in situ hybridization.