Phenol ≥99.0%, crystalline, Ultra Pure Grade

Supplier: VWR
Danger

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

0126-100G 0126-500G
97064-516CS 1747.8 USD
97064-516 97064-518
Phenol ≥99.0%, crystalline, Ultra Pure Grade
Phenol
Formula: C₆H₅OH
MW: 94.11 g/mol
Boiling Pt: 181.7
Melting Pt: 40.8
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 81 °C (closed cup)
Storage Temperature: Freezer
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1 II
Merck Index: 12,07390

Order Now

Specification Test Results

Heavy Metals (as Pb) 0.01 %
Melting Point 40-41°C
Moisture (KF) ≤ 0.5 %
pH (Saturated) @25C Reported
Purity ≥ 99.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR