Phenol : chloroform : iso-amyl alcohol (25:24:1) (v/v/v) ≥99%, Biotechnology Grade, VWR®

Supplier: VWR
Danger

Synonyms: Carbolic acid, Hydroxybenzene, Phenic acid

0883-100ML 0883-400ML
97064-692CS 1592.09 USD
97064-692 97064-694
Phenol : chloroform : iso-amyl alcohol (25:24:1) (v/v/v) ≥99%, Biotechnology Grade, VWR®
Phenol : Chloroform : iso-Amyl alcohol

Phenol:Chloroform pH 6.7/8.0 is premixed with isoamyl alcohol (25:24:1). A Phenol : Chloroform mixture is ideal for the extraction of protein from DNA preparations. Packaged at pH 6.7, this product is accompanied by a separate alkaline buffer for increasing the pH to 8.0. Doubly distilled, high purity phenol packaged under nitrogen. Safer, easier alternative to crystalline phenol.

Storage Temperature: Refrigerator
MDL Number: MFCD00133763
CAS Number: 136112-00-0
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Colorless, 2-layer liquid
DNase none detected
O.D.@ 330nm = 0.2
O.D.@ 405nm = 0.1
O.D.@ 510nm = 0.1
pH @25°C 6.5 - 6.9
Protease none detected
Purity = 99%
RNase none detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR