Chloroform : isoamyl alcohol (24:1) v/v, Biotechnology Grade
Supplier: VWR
|
Danger
|
X205-450ML
X205-950ML
97064-966EA
74.98
USD
97064-966
97064-968
Chloroform : isoamyl alcohol (24:1) v/v, Biotechnology Grade
Chloroform : isoamyl alcohol
Formula:
CHCl₃,C₅H₁₂O Storage Temperature: Ambient |
UN:
1888 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...