IPTG (isopropyl-β-D-thiogalactopyranoside), Ultrapure
Supplier: VWR
|
Warning
|
Synonyms:
(2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol
, Isopropyl Beta-D-1-thiogalactopyranoside
N679-10G
97063-282EA
683.39
USD
97063-282
IPTG (isopropyl-β-D-thiogalactopyranoside), Ultrapure
IPTG (Isopropyl-β-D-thiogalactopyranoside)
Non-animal origin (NAO) IPTG is recommended lacZ selection applications where animal derived products are prohibited. NAO IPTG is free of animal products and dioxane free. Purity is greater than 98%.
Formula:
C₉H₁₈O₅S MW: 238.31 g/mol |
MDL Number:
MFCD00063273 CAS Number: 367-93-1 |
Specification Test Results
Abs.@300nm (5%, Water) |
|
Abs.@400nm (5%, Water) |
|
Dioxane Test | Pass |
Appearance | White crystalline powder |
Melting Point | 110°C - 114°C |
Moisture (KF) |
|
pH (5%, Water)@25°C | 5 - 7 |
Purity |
|
Specific Rotation | -34.5° - -28.5° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...