G418 solution, Biotechnology Grade

Supplier: VWR

Synonyms: (2R,3S,4R,5R,6S)-5-Amino-6-(((1R,2S,3S,4R,6S)-4,6-diamino-3-(((2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-2-((R)-1-hydroxyethyl)tetrahydro-2H-pyran-3,4-diolG-418

J847-20ML
97064-358EA 70.03 USD
97064-358
G418 solution, Biotechnology Grade
G418

Selection agent for both prokaryotic and eukaryotic transfected cells


Recommended working concentration: 400 µg/ml


Mode of Action: An aminoglycoside similar to gentamycin, G418 is toxic to bacterial, yeast, higher plant and mammalian cells in addition to protozoans and helminths. Transformants survive in G418 supplemented media by expression of an aminoglycoside-modifying enzyme.

Formula: C₂₀H₄₀N₄O₁₀
MW: 496.552 g/mol
MDL Number: MFCD30541267
CAS Number: 49863-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR