Gentamicin sulfate solution for tissue culture

Supplier: VWR

Synonyms: NMD Inhibitor IIGaramycinGentiomycin CGentamicin sulfate saltGentamicin sulphate salt

E737-20ML
97062-974EA 258.24 USD
97062-974
Gentamicin sulfate solution for tissue culture
Gentamicin sulphate

15 µg/ml working concentration, binds to the 30S subunit of bacterial ribosome.


  • pH (25°C) 3.5 - 5.5
  • Specific Rotation (degrees) +107 to +121
  • Filtered through a 0.22 µM filter


Store between 2° C and 8° C

MDL Number: MFCD00270181
CAS Number: 1405-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR