Kanamycin sulfate 50 mg/mL, Ultra Pure Grade
Supplier: VWR
Synonyms:
Kanamycin Monosulphate
, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate
, Kanamycin A
, Kanamycin Monosulfate
, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulfate
Kanamycin is an aminoglycoside antibiotic that inhibits protein synthesis in gram-negative and gram-positive bacteria and in mycoplasma.
Recommended working concentration: 30 µg/mL.
Formula:
C₁₈H₃₈N₄O₁₅S MW: 582.6 g/mol |
MDL Number:
MFCD00070253 CAS Number: 25389-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...