Orthophosphoric acid 85 ACS

Supplier: VWR
Danger

Synonyms: Phosphoric acid

E582-500ML E582-50ML
97064-778CS 731.69 USD
97064-778 97064-780
Orthophosphoric acid 85 ACS
Orthophosphoric acid
Formula: H₃PO₄
MW: 98 g/mol
Boiling Pt: 158
Melting Pt: 21
Density: 1.71…1.87 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8 III
Merck Index: 14,07344

Order Now

Specification Test Results

Antimony 0.002%
APHA = 10
Arsenic = 0.0001%
Assay = 85%
Calcium = 0.002%
Chloride = 0.0003%
Heavy Metals = 0.001%
Insoluble Matter = 0.001%
Iron = 0.003%
Magnesium = 0.002%
Manganese = 0.5 ppm
Nitrate = 0.0005%
Potassium = 0.005%
Reducing Substances Pass
Sodium = 0.025%
Sulfate = 0.003%
Volatile Acids = 0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR