Methanol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Wood alcohol, Methyl alcohol, Methyl hydroxide, Hydroxy methane, Carbinol, Methylol
L13255-AU
L13255-0F
L13255-AP
AAAL13255-AUEA
59.93
USD
AAAL13255-AU
AAAL13255-0F
AAAL13255-AP
Methanol 99%
Methanol
Purity: 99%
CAS No.: 67-56-1
Molecular Formula: CH3OH
Formula Weight: 32.04
Flash Point: 11°(52°F)
Refractive Index: 1.3290
Odor: Mild
Melting Point: -98°
Boiling Point: 64.7°
Beilstein Reference: 1098229
Merck Reference: 14,5957
UN No.: 1230
MDL No.: MFCD00004595
Formula:
CH₃OH MW: 32.04 g/mol Boiling Pt: 64.6 Melting Pt: -98 Density: 0.791 Flash Pt: 11 °C (52 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004595 CAS Number: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3 II REACH: 01-2119433307-44 Merck Index: 14,5957 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...