2-Propanol ≥99.5%

Supplier: TCI America
Danger

Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol

I0164-500ML I0164-100ML
TCI0164-500MLEA 52.6 USD
TCI0164-500ML TCI0164-100ML
2-Propanol ≥99.5%
2-Propanol

[for Spectrophotometry]
CAS Number: 67-63-0
MDL Number: MFCD00011674
Molecular Formula: C3H8O
Molecular Weight: 60.10
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): 15
Specific Gravity (20/20): 0.79


Product Categories: Analytical Science, Solvents for Spectrophotometry
Other Product Names / Synonyms: Isopropanol, 2-Propanol, IPA

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.79 (20 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ef0a3050bc3fd54fa52c35e004090bd986d097f&vid=695ce94126a68bd2a2e0e9965b4f910241212a78&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR