2-Propanol ≥99.5%
Supplier: TCI America
|
Danger
|
CAS Number: 67-63-0
MDL Number: MFCD00011674
Molecular Formula: C3H8O
Molecular Weight: 60.10
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): 15
Specific Gravity (20/20): 0.79
Other Product Names / Synonyms: Isopropanol, 2-Propanol, IPA
Formula:
(CH₃)₂CHOH Boiling Pt: 82 °C Melting Pt: –89 °C Density: 0.79 (20 °C) Flash Pt: 15 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 EINECS: 200-661-7 UN: 1219 ADR: 3,II REACH: 01-2119457558-25 Merck Index: 13,05228 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...