Acetonitrile ≥99.7%, Environmental Grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

040972-M1 040972-K2 040972-K7
AA40972-M1EA 244 USD
AA40972-M1 AA40972-K2 AAA40972-K7
Acetonitrile ≥99.7%, Environmental Grade
Acetonitrile

Suitable for environmental, pesticide residue, and GC analysis


Purity: 99.7+% CAS No.: 75-05-8 Molecular Formula: CH3CN Formula Weight: 41.05 Flash Point: 5°(41°F) Physical Form: Liquid Density: 0.786 Refractive Index: 1.3440 Odor: Pungent Melting Point: -48° to -45° Boiling Point: 80-82° Beilstein Reference: 741857 Merck Reference: 14,70 UN No.: 1648 MDL No.: MFCD00001878


Suitable for environmental, pesticide residue, and GC analysis

Formula: H₃CCN
Boiling Pt: 80…82 °C
Melting Pt: –48…–45 °C
Density: 0.786
Flash Pt: 5 °C (41 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 14,70
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form Liquid
H2O 0.0005
Halogenated residue 10ng/L (GC-ECD)
ng/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR