Formaldehyde 37% in aqueous solution stabilized, Proteomics Grade

Supplier: VWR
Danger

Synonyms: MethylaldehydeMethylaldehyde solutionFormolMethanal solutionFormalinFormaline solutionParaformaldehyde aqueous solution

Stabilization: Stabilized with methanol 10.5-13 %

M134-500ML M134-200ML
97064-886CS 1249.17 USD
97064-886 97064-888
Formaldehyde 37% in aqueous solution stabilized, Proteomics Grade
Formaldehyde
Formula: CH₂O
MW: 30.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 2209
ADR: 8,III

Order Now

Specification Test Results

Acidity 0.03%
Color (APHA) = 10
Formaldehyde 37% - 37.5%
Methanol 10.5% - 13%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR