Puromycin dihydrochloride, Ultra Pure Grade
Supplier: VWR
Warning
Synonyms: (S)-2-Amino-N-((2S,3S,4R,5R)-5-(6-(Dimethylamino)-9H-purin-9-yl)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)-3-(4-methoxyphenyl)propanamide dihydrochloride
J593-25MG
97064-280EA 68.45 USD
97064-280
Puromycin dihydrochloride, Ultra Pure Grade
Puromycin dihydrochloride
A broad spectrum antibiotic that inhibits protein synthesis in both procaryotic and eucaryotic organisms
Widely used as a selection agent for cells carrying the Pac resistance gene encoding puromycin N-acetyl-transferase.