Formic acid ≥94.5%, high purity

Supplier: VWR
Danger

Synonyms: Aminic Acid, Methanoic acid

0961-500ML 0961-100ML
97064-708CS 1261.23 USD
97064-708 97064-706
Formic acid ≥94.5%, high purity
Formic acid
Formula: HCO₂H
MW: 46.03 g/mol
Boiling Pt: 100…101 °C
Melting Pt: 8.5 °C
Density: 1.22 g/cm³ (25 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8 (3),II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity = 94.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR