Gentamicin sulfate for tissue culture

Supplier: VWR
Danger

Synonyms: NMD Inhibitor IIGaramycinGentiomycin CGentamicin sulfate saltGentamicin sulphate salt

VWRV0304-100G 0304-10G 0304-5G VWRV0304-1KG VWRV0304-50G
75982-634EA 1225.35 USD
75982-634 75856-638 75856-646 97061-370 97061-372
Gentamicin sulfate for tissue culture
Gentamicin sulphate

Antibiotic that primarily targets gram-negative bacteria by binding to the 30S subunit of bacterial ribosome.


Recommended working concentration: 15 µg/ml.

Melting Pt: 218…237 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00270181
CAS Number: 1405-41-0
EINECS: 215-778-9
Merck Index: 13,04403

Order Now

Specification Test Results

Potency (on Dried Basis) Min. 590 μg/mg
Potency (as is) 0 - 9999 μg/mg
Identification (IR) Passes Test
Specific Optical Rotation (10 mg/ml) 107 - 121°
pH (25 °C, 4%) 3.5 - 5.5
Loss on Drying (110 °C; 5 mm Hg; 3 h) Max. 18.0%
Residue on Ignition Max. 1.0%
Limit of Methanol Max. 1.0%
Gentamicin C1 25 - 50%
Gentamicin C1a 10 - 35%
Gentamicin C2+C2a 25 - 55%
Gentamicin C2b+C1 25 - 50%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR