Adenine, high purity
Supplier: VWR
|
Danger
|
Synonyms:
7H-Purin-6-amine
, 6-Aminopurine
, Purin-6-amine
0183-100G
0183-50G
97061-158CS
1462.67
USD
97061-158
97061-160
Adenine, high purity
Adenine
Aápurineáderivative with essential functions for all known life forms. Required for energy metabolismáand as a component of DNA and RNA.áComponent of many culture media.á
Formula:
C₅H₅N₅ MW: 135.13 g/mol Density: 0.99741 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00041790 CAS Number: 73-24-5 EINECS: 200-796-1 UN: 2811 ADR: 6.1,III Merck Index: 13,00152 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...