KIMBERLY-CLARK PROFESSIONAL® WYPALL® Microfiber Cloths with Microban® Protection
Supplier: Janitorial Supplies
KCC83620 KCC83610
500030-637PK 17.98 USD
500030-637 500030-638 500030-639
KIMBERLY-CLARK PROFESSIONAL® WYPALL® Microfiber Cloths with Microban® Protection
Wipes Reusable Cleaning Cloths Cloths for General Applications
A 'greener' cleaning option because you can both clean and remove bacteria without using added chemicals.
Fiber structure allows cloths to pick up and trap dirt more effectively than traditional cloths. Absorbs up to eight times its weight in liquid. Color-coding helps reduce cross-contamination. Can be used wet or dry. Can be laundered up to 300 times.