Memphis™ Dyneema® Gloves, Essendant
Supplier: Janitorial Supplies
CRW9672L CRW9672XL
500012-832PR 10.87 USD
500012-832 500012-835
Memphis™ Dyneema® Gloves, Essendant
Gloves
The economical choice for solid protection against cuts and abrasions.
  • Flexible material remains breathable and lint free
  • Laundry safe and easy to clean, with no loss in cut resistance
Polyurethane palm and finger coating provides a sturdy, durable grip in both wet and dry applications.
Certifications: EN 388 (4341)