Dri-Mark® Smart Money® Pen, Essendant
Supplier: Janitorial Supplies
DRI3513B1 DRI351B1 DRI351R1 DRI351UVB
500020-567PK 12.61 USD
500020-567 500020-562 500020-566 500042-487
Dri-Mark® Smart Money® Pen, Essendant
Cash and Check Handling Supplies Counterfeit Bill Detectors
Highly effective and inexpensive tool for detecting bad bills and deterring counterfeiters from passing phony currency in the first place
  • Simply make a small mark on U.S. currency
  • Chemically sensitive ink works instantly
  • An amber mark passes the test, while a dark brown or black mark means the bill is probably counterfeit
  • Pen-style for extreme portability
Works on all new design U.S. currency (bills series 1959 and later).