Safco® Restease™ Adjustable Footrest
Supplier: Safco
Safco® Restease™
SAF2120BL
500012-420EA 63.71 USD
500012-420
Safco® Restease™ Adjustable Footrest
Ergonomic Supports Footrests
Comfort for those tired feet! Four angle adjustments from 6-25° and height adjusts from 3-1/4" to 5" for personal comfort preference
Supports feet to help reduce leg and back pressure, while integrated anti-slip rubber mat on platform keeps feet in desired position.