Swingline® Model 67 Electric Stapler

Supplier: Essendant
Swingline®
SWI06701
500025-550EA 343.29 USD
500025-550
Swingline® Model 67 Electric Stapler
Staplers Office Staplers
Everyday stapling ease you can count on

  • 100% Performance Guarantee
  • Adjustable gauge for stapling in different positions
  • Safety shut-off
  • Suction feet provide stability
  • Constructed of sturdy plastic and metal
  • Front loads a full strip (210) of standard staples
  • Seven-foot cord

Uses standard 0.25" staples.
Throat depth adjustable to 1"
Replacement staples SWI35450
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR