Kensington® SmartSockets® Color-Coded Six-Outlet Strip Surge Protector, Essendant
Supplier: Janitorial Supplies
KMW62146 KMW62147 KMW62144
500026-098EA 32.33 USD
500026-098 500026-100 500026-099
Kensington® SmartSockets® Color-Coded Six-Outlet Strip Surge Protector, Essendant
Power Supplies
Colors identify each piece of equipment for easier, simpler cord management; just match outlet colors to label colors on your equipment
  • 6-outlet unit offers quality protection
  • Power cord features a right-angle plug for easy accessibility
  • Status light
  • Unique housing is black with multi-color outlets
Noise protection up to 20 dB
Connected Equipment Warranty provides the security you expect
Basic: $10000 Limited
Premium: $25000 Limited
Strip: $15000 Limited
Certifications: UL Listed.