Kidde Pro Line™ Tri-Class Dry Chemical Fire Extinguishers
Supplier: Janitorial Supplies
KID466227 KID466112
500033-310EA 47.41 USD
500033-310 500033-312 500033-308 500033-306
Kidde Pro Line™ Tri-Class Dry Chemical Fire Extinguishers
First Aid Supplies Fire Extinguishers
Tri-Class Dry Chemical Fire Extinguishers is designed for standard commercial applications and office areas. It is effective against all three common classes of fires: A: Common combustibles—wood, paper, cloth, rubber, trash and most plastics; B: Flammable liquids—gasoline, kerosene, oil, grease, solvents and paint; C: Live electrical equipment—motors, computers, wiring and appliances (requires a non conductive extinguishing agent).
  • Seamless cylinders have chip-resistant epoxy finish
  • Reliable gauge for fast and easy pressure checks
  • Mounting bracket
More Product Information