Franklin Cleaning Technology® Compare™ Cleaner
Supplier: Essendant
Compare™
FKLF216022CT FKLF216022EA FKLF216026
500032-610CT 38.78 USD
500032-610 500032-612 500032-614
Franklin Cleaning Technology® Compare™ Cleaner
Cleaning Agents Detergents and Surface Cleansers
Triple-action, heavy-duty neutral cleaner.
  • Herbal scented with fresh fragrance
  • Low foaming, rinse-free, biodegradable
  • Contains no solvents or harsh alkalies
  • Recommended normal dilution (1:64)
For all floor surfaces not harmed by water, suitable for wood. Ideal general-purpose cleaner, mop-on floor cleaner or automatic scrubber cleaner.