Tripp Lite CAT6 Snagless Patch Cable
Supplier: Janitorial Supplies
TRPN201001BK TRPN201001BL
500044-620EA 3.27 USD
500044-620 500044-621
Tripp Lite CAT6 Snagless Patch Cable
Cables
Designed for use in CAT6 applications, this cable enables high-speed distribution of data, voice and video
High-quality copper wire and a staggered-pin plug design drastically reduce impedance and structural return loss compared to standard 100 MHz wire. Snagless design protects the connectors’ locking tabs from being damaged or snapped off during installation.