Scotch® Heat-Free 25" Laminating Machine, Essendant
Supplier: Essendant
Scotch®
MMMLS1050
500016-325EA 2807.51 USD
500016-325
Scotch® Heat-Free 25" Laminating Machine, Essendant
Laminators
Perfect lamination without electricity
  • Three easy steps: load, turn, and trim
  • Ideal for posters, banners, artwork, signs, menus, maps, and more
  • Fully compatible with inkjet, thermal, and other sensitive papers
  • Maximum document width is 25"
  • Maximum document thickness is 8.6 mil
Laminates both sides for enhanced protection, or laminates one side and applies permanent adhesive to the other for signs or labels. Laminating cartridges included (also sold separately). For documents up to 3/16" thick.
Safe, heat-free, and easy to use, with no warm-up waiting time, no fumes, and no mess.