LAVENDER™ Nitrile Exam Gloves, Kimberly-Clark
Supplier: Kimberly-Clark
KIMBERLY-CLARK® LAVENDER™
52816 52817 52820 52819 52818 52816 52819 52818 52820
89166-211CS 764.82 USD
CA89166-209 89166-211 89166-213 CA89166-211 CA89166-213 89166-205 CA89166-205 89166-207 89166-209
LAVENDER™ Nitrile Exam Gloves, Kimberly-Clark
Gloves
These latex-free, ambidextrous gloves are more economical than standard nitrile gloves and provide less hand fatigue, easier movement, and cooler hands.
  • Latex-free, powder-free, nonsterile, static dissipative
  • Ambidextrous, beaded cuffs, textured fingertips
  • Thickness: 2.8 mil (finger), 2 mil (palm), 1.6 mil (cuff)
LAVENDER™ gloves feature beaded cuffs and textured fingertips. Static dissipative in use. Powder-free. Non sterile. AQL 1.5.
Ordering information: Gloves are packaged 250 gloves (230 for X-large) per dispenser box, 10 dispenser boxes per case. Shipping case and inner packaging are labeled with product name, size, lot number, and catalog number. Environmentally responsible packaging allows space and waste reduction.