N-Methyl-2-pyrrolidone (NMP), Semi Grade for the electronics industry, VWR Chemicals BDH®
Supplier: BDH
|
Danger
|
Synonyms:
N-Methylpyrrolidinone
, N-methylpyrrolidone
, N-Methyl-gamma-butyrolactam
, N-Methyl-2-pyrrolidinone
, M-Pyrol
, N-Methyl-alpha-pyrrolidone
, 1-Methyl-2-pyrrolidone
, NMP
, Methyl Pyrrolidone
, N-Methyl-2-pyrrolidone
, N-Methyl-2-pyrrolidinone,NMP
, N-Methylpyrrolidone
, 1-Methyl-2-pyrrolidinone
10704S1G15BD
BDH2037-1GLPCS
1183.87
USD
BDH2037-1GLP
N-Methyl-2-pyrrolidone (NMP), Semi Grade for the electronics industry, VWR Chemicals BDH®
N-Methyl-2-pyrrolidone (NMP)
Formula:
C₅H₉NO MW: 99.13 g/mol Boiling Pt: 202 °C (1013 hPa) Melting Pt: –24 °C Density: 1.033 g/cm³ (20 °C) Flash Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003193 CAS Number: 872-50-4 EINECS: 212-828-1 Merck Index: 12,06197 |
Specification Test Results
Assay (CH3N(CH2)3CO) | 99.5% Min. |
Color (APHA) | 50 APHA Max. |
Residue after Ignition | 5 ppm Max. |
Water (H2O) | 0.05% Max. |
Chloride (Cl) | 1000 ppb Max. |
Phosphate (PO4) | 5000 ppb Max. |
Arsenic and Antimony | 100 ppb Max. |
Al Impurities | 100 ppb Max. |
B Impurities | 100 ppb Max. |
Cr Impurities | 100 ppb Max. |
Cu Impurities | 100 ppb Max. |
Au Impurities | 100 ppb Max. |
Fe Impurities | 100 ppb Max. |
Mg Impurities | 100 ppb Max. |
Mn Impurities | 100 ppb Max. |
Ni Impurities | 100 ppb Max. |
K Impurities | 100 ppb Max. |
Na Impurities | 100 ppb Max. |
Pb Impurities | 100 ppb Max. |
Sn Impurities | 100 ppb Max. |
Ti Impurities | 100 ppb Max. |
Zn Impurities | 100 ppb Max. |
Particles ≥1.0 µm | 10/mL Max. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...