Axygen® 384-Well PCR Plates, Corning
Supplier: Corning
PCR-384-C
10011-194CS 491.49 USD
10011-194
Axygen® 384-Well PCR Plates, Corning
Microplates PCR Plates
These plates are made of 99.9% pure virgin polypropylene and specifically designed for PCR applications
  • Max. RCF 4000 g
Optional AxyMat™ silicone sealing mat (10011-122) is autoclavable and freezable down to liquid nitrogen temperature levels. Mat is pierceable and self-sealing to prevent escape of evaporated aerosols and pathogens during normal use or storage. Alphanumeric grid system corresponds to 384-well plates.
Plates are ideal for thermal transfer applications because their ultra-thin walls accelerate heat transfer to provide shorter cycles. Certified RNase-, DNase-, and pyrogen-free to ensure purity.
Ordering information: For sealing films, including film appropriate for Real Time PCR, see 10011-121 series.