EPA method 624 Surrogate Standards, Restek
Supplier: RESTEK CORP
30026 30027
RK30026EA 42.62 USD
RK30026 RK30027
EPA method 624 Surrogate Standards, Restek
Standards Environmental Analysis Standards
Standards are packaged in purge-and-trap-grade methanol at a concentration of 2000 µg/mL in a 1 ml ampoule.
  • 2000 μg/mL in P&T methanol, 1 mL/ampoule
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).