EPA Method 527, Ultra Scientific
Supplier: Agilent
PPM-527B-1 ISM-710-1 PPM-527A-1 PPM-527C-1
ULPPM-527C-1EA 958.5 USD
ULPPM-527C-1 ULISM-710-1 ULPPM-527B-1 ULPPM-527A-1
EPA Method 527, Ultra Scientific
Standards Environmental Analysis Standards
EPA Method 527 is used to determine SOCs
The surrogate standard solution contains three analytes at 500µg/mL in actetone. Pesticide mixture #1 and #2 each contain five analytes at 500µg/mL in ethyl acetate. PDBE mixture contains 16 analytes at 500µg/mL in ethyl acetate. Volume: 1mL.