Reversible Air-Powered Stirrer, Arrow
Supplier: Arrow Fastener
G-RV
59025-122EA 1687.9 USD
59025-122
Reversible Air-Powered Stirrer, Arrow
Stirrers Overhead Stirrers
Explosion-proof reversible stirrer can be set to rotate in the direction you choose by simply connecting the air supply line to the right port for counter-clockwise rotation or the left port for clockwise rotation
Heavy-duty high-torque stirrers are recommended for batches up to 200L depending on viscosity. Constant speed (50–1000 rpm) is attained through a 7 to 1 heavy-duty gear reduction, minimizing speed fluctuations from changes in air pressure.
Unit includes a needle valve to allow variable mixing speed and a special muffler that ensures quiet operation.
Ordering information: Complete with detachable, corrosion-resistant, 30.5Lx1dia. cm (12x3/8") stainless steel shaft and 6.4dia. cm (21/2") propeller; 1.2m (4') air hose with double fittings; and 25.4Lx1.4dia. cm (10x9/16") mounting rod.