Synthware Heavy-Wall Pressure Vessel, Kemtech America
Supplier: Kemtech America
P160001 P160002 P160003 P160004 P160005 P160006 P170001 P170002 P170003 P170004 P170005 P170006
60003-766EA 78.98 USD
60003-766 60003-768 60003-770 60003-772 60003-774 60003-776 60003-792 60003-794 60003-796 60003-798 60003-800 60003-802
Synthware Heavy-Wall Pressure Vessel, Kemtech America
Reaction Vessels Reaction Vessels, Cylindrical Shape
Heavy-wall vessels are available in round bottom or cylindrical formats.
  • Internally threaded
  • Heavy wall Borosilicate construction
  • Cylindrical design
  • Protective outer coating
  • GL 15 Thread
Caution: Due to varying conditions, glass vessels cannot be guaranteed against breakage under pressure. Always work behind a safety shield.
They feature an internal thread for use with a 15 mm PTFE bushing pressure seal. The unique design provides easy access into the vessel. For improved sealing, the O-ring is positioned on the bushing above the thread. TheseMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable