Soldering Tips for PACE® Soldering Irons, Plato™, ITW Chemtronics
Supplier: ITW Chemtronics
33-7103 33-6056 33-5895 33-8143 33-1144 33-1658 SMP-2004 33-8141 33-6055 33-1146
15551-956EA 23.78 USD
15551-956 15551-932 15551-922 15552-074 15551-982 15551-960 15551-986 15551-920 15551-972 15551-962
Soldering Tips for PACE® Soldering Irons, Plato™, ITW Chemtronics
Soldering Supplies Soldering Tips
The 4.8mm (0.188") regular chisel tips (15551-932) specifically fit PACE® SP-1A, SP-2A, PS-80, ST-45, IR-70 SODR-PEN™ soldering irons.
The Extended Reach and Thermo-Drive tips can be used for the PACE® SP-2A and IR-70 soldering irons.