BioReady Gold Nanospheres, 80 nm for Passive Adsorption

Supplier: NANOCOMPOSIX, INC.
AUCR80-100M AUCR80-30M AUCR80-5M
77680-036EA 1415.5 USD
77680-036 77680-038 77680-040
BioReady Gold Nanospheres, 80 nm for Passive Adsorption
Beads Microcarriers
80 nm spherical gold nanoparticles for passive adsorption combine strong plasmonic optical properties with surface chemistry designed for passive adsorption. These nanoparticles are used in applications such as immunodiagnostics, lateral flow assays, nanomedicine, drug delivery, biosensors, and more.

  • Hydrodynamic Ø (DLS): TEM Ø + 0 to10 nm
  • Zeta potential: ≤–25 mV
  • pH of solution: 5.5 to 8.0
  • Peak wavelength (λmax): 520 to 550 nm
  • Solvent: 0.02 mM sodium citrate

Certifications: ISO 13485/cGMP.

Caution: Intended for research use only.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR