3M™ GoggleGear™ 3000 Protective Goggles
Supplier: 3M
7100325035 7100325038 7100325036 7100325034
77705-716CS 145.39 USD
77705-716 77705-720 77705-718 77705-714
3M™ GoggleGear™ 3000 Protective Goggles
Goggles
3M™ GoggleGear™ protective goggle 3000 series features a full-size design with wrap-around lens for improved field of vision, and slotted nose bridge for a more comfortable fit. Features optimized shroud design to accommodate many prescription eyewear.
  • Polycarbonate and acetate lenses absorb 99.9% of UVA, UVB, and UVC rays bet een 200 and 380 nm
  • Large profle protective goggle with wraparound lens ftting over many prescription eyewear
  • Soft, fexible shroud with slotted nose bridge design conforms to the face for added comfort
  • Indirect venting to help reduce
More Product Information