BioReady Buffers
Supplier: NANOCOMPOSIX, INC.
The same high quality conjugation reaction buffers, block buffer, and conjugate diluent for covalent conjugation found in our high sensitivity gold conjugation kit, now available in catalog and bulk volumes.
- Convenient one-stop shopping for conjugation buffers and nanoparticles
- Save time on formulating buffers and writing SOPs
- Ensure consistent quality and supply for your conjugation
BioReady Block Covalent Buffer
4 mM sodium tetraborate, 1% BSA, 0.05% sodium azide, pH 8.2
BioReady Running Buffer
1X PBS, 1% Tween 20, pH 7.4
BioReady Sodium Phosphate Reaction Buffer
5mM sodium phosphate, 0.5% PEG20K, pH 7.4
BioReady Conjugate Diluent
0.5X PBS, 0.5% BSA, 0.5% casein, 1% Tween 20, 0.05% sodium azide, pH 8.0
BioReady Potassium Phosphate Reaction Buffer
5 mM potassium phosphate, 0.5% PEG20K, pH 7.4
BioReady PBS Reaction Buffer
0.01X PBS, 0.5% PEG20K, pH 7.4
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...