Anti-BRCA1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43734-100UG A43734-50UG
77706-410EA 334.95 USD
77706-410 77706-411
Anti-BRCA1 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: BRCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR