Anti-HDC Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45807-100UL A45807-50UL
77710-088EA 334.95 USD
77710-088 77710-089
Anti-HDC Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, IHC:1:5-1:20

Type:
Antigen: HDC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR