Anti-IFNG Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45779-100UG A45779-50UG
77689-196EA 334.95 USD
77689-196 77710-038
Anti-IFNG Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000

Type:
Antigen: IFNG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR