Titanium, Ti 4-2.5-1.5, Grade 38, R54250, 65g, ARMI | MBH, LGC Standards

Supplier: LGC Standards

IARM-TI42515-18-C IARM-TI42515-18-G IARM-TI42515-18-X
77693-623EA 854.65 USD
77693-623 77693-624 77693-625
Titanium, Ti 4-2.5-1.5, Grade 38, R54250, 65g, ARMI | MBH, LGC Standards
Standards Alloys and Metal Ore Standards

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR