Anti-NEUROD6 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A64383-100UG A64383-50UG
77723-919EA 334.95 USD
77723-919 77723-920
Anti-NEUROD6 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000

Type:
Antigen: NEUROD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR